siRNA Details
| Gene Symbol: Rhod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007485.4 | NP_031511.1 | 5 | 657 | ||
| GenBank mRNA | AK008880.1 | - | 5 | 658 | ||
| BC005652.1 | - | 4 | 1024 | |||
| BC047989.1 | - | 5 | 666 | 6333308 | ||
| D89821.1 | - | 668 | ||||
| X84325.1 | - | 5 | 584 | |||

Rhod
Genomic Map