siRNA Details
| Gene Symbol: Arnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037737.2 | NP_001032826.1 | 7 | 756 | ||
| NM_009709.4 | NP_033839.2 | 6 | 711 | |||
| GenBank mRNA | AK028546.1 | - | 6 | 698 | ||
| AK040475.1 | - | 7 | 758 | |||
| AK141362.1 | - | 7 | 627 | |||
| AK145415.1 | - | 6 | 666 | |||
| AK152473.1 | - | 7 | 738 | |||
| AK153355.1 | - | 7 | 738 | |||
| AK220458.1 | - | 3 | 887 | |||
| BC012870.1 | - | 7 | 718 | 3964932 | ||
| U10325.1 | - | 7 | 620 | |||
| U14333.1 | - | 6 | 549 | |||

Arnt
Genomic Map