siRNA Details
| Gene Symbol: Asah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019734.2 | NP_062708.1 | 4 | 354 | ||
| GenBank mRNA | AF157500.1 | - | 4 | 284 | ||
| AF352179.1 | - | 4 | 296 | |||
| AK075666.1 | - | 4 | 269 | |||
| AK075919.1 | - | 4 | 308 | |||
| AK077721.1 | - | 4 | 307 | |||
| AK090314.1 | - | 4 | 304 | |||
| AK150567.1 | - | 4 | 267 | |||
| AK151208.1 | - | 4 | 355 | |||
| AK151312.1 | - | 4 | 267 | |||
| AK151478.1 | - | 4 | 345 | |||
| AK152305.1 | - | 4 | 306 | |||
| AK152811.1 | - | 4 | 267 | |||
| AK152920.1 | - | 4 | 314 | |||
| AK153101.1 | - | 4 | 314 | |||
| AK153297.1 | - | 4 | 268 | |||
| AK159556.1 | - | 4 | 276 | |||
| AK166757.1 | - | 4 | 286 | |||
| BC003204.1 | - | 4 | 274 | 3587042 | ||
| BY160973.1 | - | 4 | 355 | |||

Asah1
Genomic Map