siRNA Details
| Gene Symbol: Rab27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301230.1 | NP_001288159.1 | 4, 5 | 615 | ||
| NM_001301232.1 | NP_001288161.1 | 3, 4 | 508 | |||
| NM_023635.6 | NP_076124.1 | 3, 4 | 466 | |||
| GenBank mRNA | AB046693.1 | - | 2, 3 | 264 | ||
| AB120511.1 | - | 2, 3 | 242 | |||
| AK010373.1 | - | 3, 4 | 384 | |||
| AK030529.1 | - | 3, 4 | 413 | |||
| BC009656.1 | - | 3, 4 | 419 | 3590089 | ||
| CV784419.1 | - | 3, 4 | 377 | |||

Rab27a
Genomic Map