siRNA Details
| Gene Symbol: Zfhx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007496.2 | NP_031522.2 | 2 | 1658 | ||
| GenBank mRNA | D26046.1 | - | 2 | 1655 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfhx3| Gene Symbol: Zfhx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007496.2 | NP_031522.2 | 2 | 1658 | ||
| GenBank mRNA | D26046.1 | - | 2 | 1655 | ||