siRNA Details
| Gene Symbol: Atm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007499.2 | NP_031525.2 | 16 | 2545 | ||
| GenBank mRNA | BC138525.1 | - | 15 | 2447 | 8861545 | |
| U43678.1 | - | 15 | 2447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atm| Gene Symbol: Atm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007499.2 | NP_031525.2 | 16 | 2545 | ||
| GenBank mRNA | BC138525.1 | - | 15 | 2447 | 8861545 | |
| U43678.1 | - | 15 | 2447 | |||