siRNA Details
| Gene Symbol: GSTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000849.4 | NP_000840.2 | 9 | 906 | ||
| NR_024537.1 | 8 | 830 | ||||
| GenBank mRNA | AB209816.1 | - | 8 | 615 | ||
| AK304300.1 | - | 7 | 558 | |||
| AK313444.1 | - | 8 | 624 | |||
| BC000088.2 | - | 8 | 611 | 3509784 | ||
| BC008790.2 | - | 9 | 906 | 3630923 | ||
| BT019945.1 | - | 8 | 596 | |||
| CR542108.1 | - | 8 | 596 | |||
| J05459.1 | - | 8 | 613 | |||

GSTM3
Genomic Map