siRNA Details
| Gene Symbol: Atp6v0d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013477.3 | NP_038505.2 | 6 | 924 | ||
| GenBank mRNA | AB088408.1 | - | 6 | 813 | ||
| AK083372.1 | - | 6 | 925 | |||
| AK133921.1 | - | 6 | 933 | |||
| AK171515.1 | - | 6 | 894 | |||
| BC011075.1 | - | 6 | 815 | 3662404 | ||
| U13840.1 | - | 6 | 813 | |||
| U21549.1 | - | 6 | 796 | |||

Atp6v0d1
Genomic Map