siRNA Details
| Gene Symbol: Atp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109757.2 | NP_001103227.1 | 16 | 3352 | ||
| NM_009726.5 | NP_033856.3 | 16 | 3349 | |||
| GenBank mRNA | AB007134.1 | - | 15 | 3188 | ||
| BC141395.1 | - | 16 | 3230 | 9056008 | ||
| U03434.1 | - | 16 | 3257 | |||
| U03736.1 | - | 15 | 3097 | |||
| U71091.1 | - | 16 | 3253 | |||

Atp7a
Genomic Map