siRNA Details
| Gene Symbol: Atp7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109757.2 | NP_001103227.1 | 8 | 2075 | ||
| NM_009726.5 | NP_033856.3 | 8 | 2072 | |||
| GenBank mRNA | AB007134.1 | - | 7 | 1911 | ||
| BC141395.1 | - | 8 | 1953 | 9056008 | ||
| U03434.1 | - | 8 | 1980 | |||
| U03736.1 | - | 7 | 1820 | |||
| U71091.1 | - | 8 | 1976 | |||

Atp7a
Genomic Map