siRNA Details
| Gene Symbol: Atp10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009728.2 | NP_033858.2 | 5 | 1221 | ||
| GenBank mRNA | AF011337.1 | - | 5 | 924 | ||
| AF156549.1 | - | 5 | 1091 | |||
| AK132174.1 | - | 5 | 1178 | |||
| AK172149.1 | - | 5 | 1142 | |||
| AK172972.1 | - | 5 | 1007 | |||
| BC138356.1 | - | 5 | 1025 | 8861376 | ||
| BC138357.1 | - | 5 | 1025 | 8861377 | ||

Atp10a
Genomic Map