siRNA Details
| Gene Symbol: Slc7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301424.1 | NP_001288353.1 | 10 | 1546 | ||
| NM_007513.4 | NP_031539.3 | 10 | 1662 | |||
| GenBank mRNA | AK139342.1 | - | 10 | 1638 | ||
| AK147991.1 | - | 10 | 1571 | |||
| AK163202.1 | - | 10 | 1546 | |||
| BC068234.1 | - | 10 | 1553 | |||
| BC145779.1 | - | 9 | 1467 | 40131082 | ||
| BC145781.1 | - | 9 | 1467 | 40131084 | ||
| JN226407.1 | - | 8 | 1382 | |||
| JN226408.1 | - | 8 | 1382 | |||
| JN226410.1 | - | 8 | 1382 | |||
| M26687.1 | - | 10 | 1580 | |||

Slc7a1
Genomic Map