siRNA Details
| Gene Symbol: Slc7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077593.1 | - | 3 | 615 | ||
| AK141558.1 | - | 3 | 646 | |||
| AK165931.1 | - | 3 | 612 | |||
| BC050195.1 | - | 3 | 591 | 30010078 | ||
| U70859.1 | - | 4 | 749 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a3| Gene Symbol: Slc7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077593.1 | - | 3 | 615 | ||
| AK141558.1 | - | 3 | 646 | |||
| AK165931.1 | - | 3 | 612 | |||
| BC050195.1 | - | 3 | 591 | 30010078 | ||
| U70859.1 | - | 4 | 749 | |||