siRNA Details
| Gene Symbol: Pcdh15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142742.1 | NP_001136214.1 | 3, 4 | 525 | ||
| NM_001142746.1 | NP_001136218.1 | 3, 4 | 525 | |||
| NM_001142747.1 | NP_001136219.1 | 3, 4 | 525 | |||
| NM_023115.3 | NP_075604.2 | 3, 4 | 525 | |||
| GenBank mRNA | AF281899.1 | - | 3, 4 | 509 | ||
| AK139154.1 | - | 3, 4 | 450 | |||
| DQ354405.1 | - | 3, 4 | 509 | |||
| DQ354413.1 | - | 3, 4 | 509 | |||
| DQ354416.1 | - | 3, 4 | 450 | |||

Pcdh15
Genomic Map