siRNA Details
| Gene Symbol: Bcap29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164090.1 | NP_001157562.1 | 4 | 285 | ||
| NM_007530.3 | NP_031556.1 | 3 | 448 | |||
| GenBank mRNA | AK090068.1 | - | 4 | 262 | ||
| AK144754.1 | - | 4 | 259 | |||
| AK218818.1 | - | 3 | 186 | |||
| BC003303.1 | - | 3 | 352 | 3584161 | ||
| BC021661.1 | - | 3 | 459 | 2646883 | ||
| X78684.1 | - | 3 | 260 | |||

Bcap29
Genomic Map