siRNA Details
| Gene Symbol: Bckdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199195.1 | NP_954665.1 | 10 | 1022 | ||
| GenBank mRNA | AK089921.1 | - | 10 | 1103 | ||
| AK156264.1 | - | 10 | 1101 | |||
| AK164531.1 | - | 3 | 165 | |||
| BC064099.1 | - | 10 | 1022 | 1246285 | ||
| L16992.1 | - | 10 | 1094 | |||

Bckdhb
Genomic Map