siRNA Details
| Gene Symbol: Bcl2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009742.3 | NP_033872.1 | 1, 2 | 544 | ||
| GenBank mRNA | L16462.1 | - | 1, 2 | 544 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcl2a1a| Gene Symbol: Bcl2a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009742.3 | NP_033872.1 | 1, 2 | 544 | ||
| GenBank mRNA | L16462.1 | - | 1, 2 | 544 | ||