siRNA Details
| Gene Symbol: Bcl7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009745.2 | NP_033875.2 | 3 | 388 | ||
| GenBank mRNA | AJ011145.1 | - | 3 | 268 | ||
| AK131643.1 | - | 3 | 284 | |||
| AK155076.1 | - | 3 | 270 | |||
| AK219150.1 | - | 1 | 52 | |||
| BC011471.1 | - | 3 | 365 | 3660754 | ||

Bcl7b
Genomic Map