siRNA Details
| Gene Symbol: Bet1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009748.2 | NP_033878.1 | 2 | 262 | ||
| GenBank mRNA | AF007552.1 | - | 2 | 250 | ||
| AK053940.1 | - | 2 | 284 | |||
| AK078796.1 | - | 2 | 258 | |||
| BC005572.1 | - | 2 | 262 | 3967968 | ||

Bet1
Genomic Map