siRNA Details
| Gene Symbol: Bex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009749.2 | NP_033879.1 | 3 | 455 | ||
| GenBank mRNA | AF097439.1 | - | 3 | 329 | ||
| AK132005.1 | - | 3 | 342 | |||
| BC027529.1 | - | 3 | 329 | 5058536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bex2| Gene Symbol: Bex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009749.2 | NP_033879.1 | 3 | 455 | ||
| GenBank mRNA | AF097439.1 | - | 3 | 329 | ||
| AK132005.1 | - | 3 | 342 | |||
| BC027529.1 | - | 3 | 329 | 5058536 | ||