siRNA Details
| Gene Symbol: Ngfrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110233.1 | NP_001103703.1 | 3 | 453 | ||
| NM_001110234.1 | NP_001103704.1 | 3 | 412 | |||
| NM_009750.2 | NP_033880.1 | 3 | 458 | |||
| GenBank mRNA | AF097440.1 | - | 3 | 379 | ||
| AF187066.1 | - | 3 | 384 | |||
| AK003294.1 | - | 3 | 366 | |||
| AK004531.1 | - | 3 | 388 | |||
| AK010500.1 | - | 3 | 385 | |||
| AV574205.1 | - | 3 | 365 | |||
| AV575702.1 | - | 3 | 459 | |||
| BC027815.1 | - | 3 | 384 | 5250429 | ||
| CB055608.1 | - | 3 | 311 | |||
| CF618290.1 | - | 3 | 402 | |||

Ngfrap1
Genomic Map