siRNA Details
| Gene Symbol: Bgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007542.4 | NP_031568.2 | 7 | 941 | ||
| GenBank mRNA | AK135496.1 | - | 7 | 942 | ||
| AK155973.1 | - | 7 | 942 | |||
| AK164867.1 | - | 7 | 942 | |||
| AK171870.1 | - | 7 | 942 | |||
| BC005452.1 | - | 7 | 935 | 3594469 | ||
| BC019502.1 | - | 7 | 944 | 4237268 | ||
| BC052857.1 | - | 7 | 926 | 30061850 | ||
| L20276.1 | - | 7 | 941 | |||
| X53928.1 | - | 1036 | ||||

Bgn
Genomic Map