siRNA Details
| Gene Symbol: Bicd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112796.2 | NP_001106267.1 | 5 | 2297 | ||
| NM_009753.4 | NP_033883.2 | 5 | 2297 | |||
| GenBank mRNA | AK033407.1 | - | 2 | 914 | ||
| AK045975.1 | - | 5 | 2216 | |||
| AK051379.1 | - | 5 | 2299 | |||
| AK082523.1 | - | 5 | 2226 | |||
| AK220497.1 | - | 5 | 2265 | |||
| BC016192.1 | - | 5 | 2000 | 4483840 | ||
| U90029.1 | - | 2 | 1164 | |||

Bicd1
Genomic Map