siRNA Details
| Gene Symbol: Bicd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112796.2 | NP_001106267.1 | 3, 4 | 1026 | ||
| NM_009753.4 | NP_033883.2 | 3, 4 | 1026 | |||
| GenBank mRNA | AJ288054.1 | - | 2, 3 | 380 | ||
| AK043650.1 | - | 3, 4 | 937 | |||
| AK045975.1 | - | 3, 4 | 945 | |||
| AK051379.1 | - | 3, 4 | 1028 | |||
| AK051718.1 | - | 3, 4 | 960 | |||
| AK082523.1 | - | 3, 4 | 955 | |||
| AK220497.1 | - | 3, 4 | 994 | |||
| BC016192.1 | - | 3, 4 | 729 | 4483840 | ||

Bicd1
Genomic Map