siRNA Details
| Gene Symbol: Prdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133503.1 | - | 7 | 2483 | ||
| AK149344.1 | - | 7 | 2374 | |||
| BC129801.1 | - | 7 | 2493 | |||
| JX154081.1 | - | 7 | 2316 | |||
| U08185.1 | - | 8 | 2633 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prdm1| Gene Symbol: Prdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133503.1 | - | 7 | 2483 | ||
| AK149344.1 | - | 7 | 2374 | |||
| BC129801.1 | - | 7 | 2493 | |||
| JX154081.1 | - | 7 | 2316 | |||
| U08185.1 | - | 8 | 2633 | |||