siRNA Details
| Gene Symbol: Bmpr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277216.1 | NP_001264145.1 | 10, 11 | 1426 | ||
| NM_001277217.1 | NP_001264146.1 | 10, 11 | 1452 | |||
| NM_001277218.1 | NP_001264147.1 | 9, 10 | 1465 | |||
| NM_001277220.1 | NP_001264149.1 | 10, 11 | 1600 | |||
| NM_007560.4 | NP_031586.1 | 9, 10 | 1619 | |||
| GenBank mRNA | AK086130.1 | - | 10, 11 | 1412 | ||
| AK140158.1 | - | 10, 11 | 1524 | |||
| AK162844.1 | - | 9, 10 | 1300 | |||
| BC065106.1 | - | 9, 10 | 1400 | 30357130 | ||
| BC065143.1 | - | 9, 10 | 1606 | 30629345 | ||
| BC138638.1 | - | 9, 10 | 1346 | 8861658 | ||
| BC138640.1 | - | 9, 10 | 1346 | 8861660 | ||
| Z23143.1 | - | 10, 11 | 1444 | |||

Bmpr1b
Genomic Map