siRNA Details
| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007561.4 | NP_031587.1 | 12, 13 | 3237 | ||
| GenBank mRNA | AF003942.1 | - | 12, 13 | 3237 | ||
| AK138715.1 | - | 2, 3 | 1365 | |||
| AK148180.1 | - | 7, 8 | 2055 | |||
| U78048.1 | - | 12, 13 | 2879 | |||

Bmpr2
Genomic Map