siRNA Details
| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007561.4 | NP_031587.1 | 6 | 1158 | ||
| GenBank mRNA | AF003942.1 | - | 6 | 1158 | ||
| AK149385.1 | - | 2039 | ||||
| U78048.1 | - | 6 | 800 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmpr2| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007561.4 | NP_031587.1 | 6 | 1158 | ||
| GenBank mRNA | AF003942.1 | - | 6 | 1158 | ||
| AK149385.1 | - | 2039 | ||||
| U78048.1 | - | 6 | 800 | |||