siRNA Details
| Gene Symbol: Bmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009759.4 | NP_033889.2 | 15 | 1553 | ||
| GenBank mRNA | AF012104.1 | - | 14 | 1406 | ||
| AK036707.1 | - | 14 | 1412 | |||
| AK040936.1 | - | 15 | 1554 | |||
| BC138104.1 | - | 15 | 1430 | 8861123 | ||
| BC138105.1 | - | 15 | 1430 | 8861124 | ||
| U88091.1 | - | 15 | 1539 | |||

Bmx
Genomic Map