siRNA Details
| Gene Symbol: Bnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008238.3 | NP_001008239.1 | 8 | 1060 | ||
| NM_016787.4 | NP_058067.2 | 8 | 1060 | |||
| GenBank mRNA | AF035207.1 | - | 8 | 912 | ||
| AF400107.1 | - | 7 | 800 | |||
| AK014659.1 | - | 8 | 1012 | |||
| AK054288.1 | - | 8 | 1026 | |||
| AK159745.1 | - | 8 | 1023 | |||
| AK161116.1 | - | 8 | 1013 | |||
| AK169609.1 | - | 8 | 1045 | |||
| AY591757.1 | - | 7 | 779 | |||
| AY591758.1 | - | 7 | 779 | |||
| BC012670.1 | - | 8 | 1019 | 4037627 | ||

Bnip2
Genomic Map