siRNA Details
| Gene Symbol: Bpgm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007563.4 | NP_031589.1 | 2 | 546 | ||
| GenBank mRNA | AK006315.1 | - | 2 | 598 | ||
| AK037239.1 | - | 2 | 547 | |||
| AK043412.1 | - | 2 | 584 | |||
| AK078119.1 | - | 2 | 547 | |||
| AK171685.1 | - | 2 | 547 | |||
| BC004589.1 | - | 2 | 571 | 3587820 | ||
| X13586.1 | - | 1 | 439 | |||

Bpgm
Genomic Map