siRNA Details
| Gene Symbol: Brca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009764.3 | NP_033894.3 | 10 | 3670 | ||
| GenBank mRNA | AB221610.1 | - | 1 | 899 | ||
| BC068303.1 | - | 10 | 3506 | 30431022 | ||
| U31625.1 | - | 10 | 3527 | |||
| U32446.1 | - | 10 | 3580 | |||
| U35641.1 | - | 10 | 3563 | |||
| U36475.1 | - | 9 | 3490 | |||
| U68174.1 | - | 10 | 3574 | |||

Brca1
Genomic Map