siRNA Details
| Gene Symbol: Brca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009764.3 | NP_033894.3 | 10 | 1309 | ||
| GenBank mRNA | AB221611.1 | - | 9 | 1103 | ||
| AK086173.1 | - | 9 | 1222 | |||
| AK144702.1 | - | 10 | 1228 | |||
| BC068303.1 | - | 10 | 1145 | 30431022 | ||
| U31625.1 | - | 10 | 1166 | |||
| U32446.1 | - | 10 | 1219 | |||
| U35641.1 | - | 10 | 1202 | |||
| U36475.1 | - | 9 | 1129 | |||
| U68174.1 | - | 10 | 1213 | |||

Brca1
Genomic Map