siRNA Details
| Gene Symbol: Birc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007566.3 | NP_031592.3 | 17 | 4195 | ||
| GenBank mRNA | AK134254.1 | - | 2 | 115 | ||
| Y17267.1 | - | 17 | 4196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Birc6| Gene Symbol: Birc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007566.3 | NP_031592.3 | 17 | 4195 | ||
| GenBank mRNA | AK134254.1 | - | 2 | 115 | ||
| Y17267.1 | - | 17 | 4196 | |||