siRNA Details
| Gene Symbol: Bsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077184.1 | NP_001070652.1 | 5 | 696 | ||
| NM_009768.2 | NP_033898.1 | 6 | 1044 | |||
| GenBank mRNA | AK002332.1 | - | 5 | 548 | ||
| AK011364.1 | - | 5 | 547 | |||
| AK147225.1 | - | 5 | 5370 | |||
| AY089967.1 | - | 6 | 902 | |||
| BC010270.1 | - | 5 | 539 | 3589236 | ||
| D00611.1 | - | 5 | 546 | |||
| S63813.1 | - | 5 | 546 | |||
| Y16256.1 | - | 5 | 546 | |||
| Y16257.1 | - | 4 | 542 | |||
| Y16258.1 | - | 4 | 488 | |||

Bsg
Genomic Map