siRNA Details
| Gene Symbol: Btg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297747.1 | NP_001284676.1 | 1, 2 | 204 | ||
| NM_009770.3 | NP_033900.1 | 2, 3 | 400 | |||
| GenBank mRNA | AK078652.1 | - | 103 | |||
| AK144415.1 | - | 2, 3 | 351 | |||
| BC012705.1 | - | 2, 3 | 339 | 3980599 | ||
| BC094027.1 | - | 2, 3 | 355 | 4457150 | ||
| BC147656.1 | - | 1, 2 | 204 | 9087445 | ||
| BC147662.1 | - | 1, 2 | 204 | 9087451 | ||
| CK623612.1 | - | 2, 3 | 400 | |||
| D83745.1 | - | 2, 3 | 357 | |||
| Z72000.1 | - | 2, 3 | 375 | |||

Btg3
Genomic Map