siRNA Details
| Gene Symbol: Btn1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013483.3 | NP_038511.1 | 8 | 1170 | ||
| GenBank mRNA | AK142596.1 | - | 8 | 1077 | ||
| AK145168.1 | - | 8 | 1171 | |||
| BC011497.1 | - | 8 | 1144 | 3583827 | ||
| BC031459.1 | - | 7 | 1068 | 3582574 | ||
| S80642.1 | - | 6 | 896 | |||

Btn1a1
Genomic Map