siRNA Details
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009774.3 | NP_033904.2 | 7 | 854 | ||
GenBank mRNA | AF149822.1 | - | 6 | 790 | ||
AK012744.1 | - | 2 | 58 | |||
AK088534.1 | - | 7 | 855 | |||
AK180056.1 | - | 1 | 48 | |||
AK201702.1 | - | 1 | 48 | |||
AK203218.1 | - | 1 | 48 | |||
AK217256.1 | - | 1 | 41 | |||
BC025089.1 | - | 7 | 866 | 3586753 | ||
BC086790.1 | - | 5 | 2819 | |||
U67327.1 | - | 7 | 841 |
