siRNA Details
| Gene Symbol: Bub3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009774.3 | NP_033904.2 | 3 | 314 | ||
| GenBank mRNA | AF149822.1 | - | 2 | 250 | ||
| AK088534.1 | - | 3 | 315 | |||
| AK190470.1 | - | 2 | 116 | |||
| BC025089.1 | - | 3 | 326 | 3586753 | ||
| BC086790.1 | - | 2 | 553 | |||
| U67327.1 | - | 3 | 301 | |||

Bub3
Genomic Map