siRNA Details
| Gene Symbol: Bub3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009774.3 | NP_033904.2 | 3 | 293 | ||
| GenBank mRNA | AF149822.1 | - | 2 | 229 | ||
| AK088534.1 | - | 3 | 294 | |||
| AK190470.1 | - | 2 | 95 | |||
| BC025089.1 | - | 3 | 305 | 3586753 | ||
| BC086790.1 | - | 2 | 532 | |||

Bub3
Genomic Map