siRNA Details
| Gene Symbol: C1qa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007572.2 | NP_031598.2 | 3 | 755 | ||
| GenBank mRNA | AK002655.1 | - | 3 | 681 | ||
| AK136737.1 | - | 3 | 681 | |||
| AK150771.1 | - | 3 | 681 | |||
| AK159210.1 | - | 3 | 681 | |||
| AK159343.1 | - | 3 | 676 | |||
| BC002086.1 | - | 3 | 714 | 3592169 | ||
| BC020096.1 | - | 2 | 676 | |||
| BY698724.1 | - | 1 | 66 | |||

C1qa
Genomic Map