siRNA Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013484.2 | NP_038512.2 | 15 | 1973 | ||
| GenBank mRNA | AK146812.1 | - | 15 | 1974 | ||
| AK190654.1 | - | 3 | 231 | |||
| AK207402.1 | - | 2 | 65 | |||
| BC011086.1 | - | 15 | 1936 | 3992881 | ||
| M57891.1 | - | 15 | 1920 | |||

C2
Genomic Map