siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016704.2 | NP_057913.2 | 3 | 292 | ||
| GenBank mRNA | AF184900.1 | - | 3 | 293 | ||
| AK041057.1 | - | 3 | 302 | |||
| BB222000.2 | - | 2 | 44 | |||
| BC011251.1 | - | 3 | 281 | 4196330 | ||
| BY088427.1 | - | 3 | 289 | |||
| BY105449.1 | - | 3 | 293 | |||

C6
Genomic Map