siRNA Details
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016704.2 | NP_057913.2 | 10 | 1575 | ||
GenBank mRNA | AF184900.1 | - | 10 | 1576 | ||
BC011251.1 | - | 10 | 1564 | 4196330 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016704.2 | NP_057913.2 | 10 | 1575 | ||
GenBank mRNA | AF184900.1 | - | 10 | 1576 | ||
BC011251.1 | - | 10 | 1564 | 4196330 |