siRNA Details
| Gene Symbol: Cacna1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB086123.1 | - | 48 | 6167 | ||
| AJ437291.1 | - | 46 | 6044 | |||
| AJ437292.1 | - | 46 | 6165 | |||
| AK018426.1 | - | 1 | 16 | |||
| AK032552.1 | - | 2 | 988 | |||
| AK147415.1 | - | 47 | 7635 | |||
| BC058783.1 | - | 16 | 2393 | |||

Cacna1d
Genomic Map