siRNA Details
| Gene Symbol: Cacnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252533.1 | NP_001239462.1 | 12 | 1374 | ||
| NM_023116.4 | NP_075605.1 | 13 | 1505 | |||
| GenBank mRNA | AK034054.1 | - | 13 | 1506 | ||
| AM259383.2 | - | 12 | 1374 | |||
| BC109156.1 | - | 13 | 1437 | 40047524 | ||
| BC109157.1 | - | 13 | 1437 | 40047531 | ||
| KJ789960.1 | - | 12 | 1093 | |||
| L20343.1 | - | 12 | 1284 | |||

Cacnb2
Genomic Map