siRNA Details
| Gene Symbol: Camk2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039138.2 | NP_001034227.1 | 15 | 1205 | ||
| NM_001039139.2 | NP_001034228.1 | 14 | 1136 | |||
| NM_178597.5 | NP_848712.2 | 16 | 1238 | |||
| GenBank mRNA | AF395884.1 | - | 16 | 1115 | ||
| AK031213.1 | - | 14 | 1099 | |||
| AK078311.1 | - | 15 | 1150 | |||
| AK081969.1 | - | 14 | 1608 | |||
| AK154764.1 | - | 15 | 1134 | |||
| AK181156.1 | - | 2 | 102 | |||
| AK185115.1 | - | 2 | 102 | |||
| BC019162.1 | - | 15 | 1157 | 3675224 | ||
| BC025597.1 | - | 14 | 1090 | 5342641 | ||

Camk2g
Genomic Map