siRNA Details
| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009793.3 | NP_033923.2 | 11 | 1422 | ||
| GenBank mRNA | AK030219.1 | - | 11 | 1306 | ||
| AK043114.1 | - | 11 | 1132 | |||
| BC070420.1 | - | 11 | 1303 | 5718423 | ||
| J03057.1 | - | 3 | 325 | |||
| M16206.1 | - | 2 | 125 | |||
| M64266.1 | - | 3 | 373 | |||
| X58995.1 | - | 11 | 1112 | |||

Camk4
Genomic Map