siRNA Details
| Gene Symbol: Capns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009795.3 | NP_033925.2 | 9 | 731 | ||
| GenBank mRNA | AK028092.1 | - | 7 | 477 | ||
| AK132819.1 | - | 8 | 541 | |||
| BC018352.1 | - | 9 | 725 | 3156021 | ||
| BC090988.1 | - | 9 | 731 | 30246016 | ||

Capns1
Genomic Map