siRNA Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301250.1 | NP_001288179.1 | 8 | 1090 | ||
| NM_007602.4 | NP_031628.1 | 8 | 1254 | |||
| GenBank mRNA | AK155179.1 | - | 8 | 1232 | ||
| AK164293.1 | - | 8 | 1164 | |||
| AK170034.1 | - | 8 | 1191 | |||
| BC014767.1 | - | 8 | 1082 | 4458488 | ||
| U85020.1 | - | 8 | 1173 | |||
| Y10656.1 | - | 8 | 1186 | |||

Capn5
Genomic Map